SY3H3D67D7F2 Synthonix Corporation

4-Ethynyl-3-fluorobenzoic acid

  • MDL number MFCD30329071
  • Cas Number : 1862427-77-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Yellow solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/50773.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/50773.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart