SY3H3D67D7FC Synthonix Corporation

Ethyl 5-chlorothiazolo[5,4-b]pyridine-2-carboxylate

  • Cas Number : 1202075-71-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/51111.html

Safety Information

MSDS https://synthonix.com/51111.html
Availability details for
Quantity
  • Availability
  • Add to Cart