SY3H3D67D800 Synthonix Corporation

1-Ethynyl-2-methoxy-4-nitrobenzene

  • MDL number MFCD11840985
  • Cas Number : 1135283-10-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/51212.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/51212.html
pictograms GHS07
hstatements H302, H315, H319, H332, H335
Availability details for
Quantity
  • Availability
  • Add to Cart