SY3H3D67D801 Synthonix Corporation

Ethyl 5-bromo-2-(2,2,2-trifluoroacetamido)thiophene-3-carboxylate

  • MDL number MFCD30536240
  • Cas Number : 2044706-51-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Green Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/51219.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/51219.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart