SY3H3D67D810 Synthonix Corporation

Ethyl 7-bromo-[1,2,4]triazolo[1,5-a]pyridine-2-carboxylate

  • Cas Number : 1380331-36-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/51436.html

Safety Information

MSDS https://synthonix.com/51436.html
Availability details for
Quantity
  • Availability
  • Add to Cart