SY3H3D67D812 Synthonix Corporation

Ethyl 7-chloropyrazolo[1,5-a]pyrimidine-5-carboxylate

  • MDL number MFCD13151991
  • Cas Number : 61098-37-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form Pale-yellow to Yellow-brown Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/51439.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/51439.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart