SY3H3D67D815 Synthonix Corporation

Ethyl 4-hydroxy-6-(trifluoromethyl)nicotinate

  • Cas Number : 1269292-28-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/51483.html

Safety Information

MSDS https://synthonix.com/51483.html
Availability details for
Quantity
  • Availability
  • Add to Cart