SY3H3D67D816 Synthonix Corporation

Ethyl 2-bromo-5-nitrobenzoate

  • Cas Number : 208176-31-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/51484.html

Safety Information

MSDS https://synthonix.com/51484.html
Availability details for
Quantity
  • Availability
  • Add to Cart