SY3H3D67D817 Synthonix Corporation

Ethyl 3-bromo-5-nitrobenzoate

  • MDL number MFCD09258760
  • Cas Number : 690260-94-7
  • Molecular Weight : 274.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl 3-bromo-5-nitrobenzoate
Inchi Code 1S/C9H8BrNO4/c1-2-15-9(12)6-3-7(10)5-8(4-6)11(13)14/h3-5H,2H2,1H3
Inchi Key PUBVWGDKFHIPRR-UHFFFAOYSA-N

Description

Purity 98%
Storage temp. Refrigerator
Physical Form Off-white to Pale-yellow to Yellow-brown Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/51485.html

Safety Information

signalword Warning
pcodes P261,P305,P351,P338
MSDS https://synthonix.com/51485.html
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart