SY3H3D67D820 Synthonix Corporation

Ethyl 2-bromo-4-fluorobenzoate

  • MDL number MFCD06204924
  • Cas Number : 651341-68-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Colorless to light yellow to pale-brown liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/51574.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/51574.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart