SY3H3D67D828 Synthonix Corporation

Ethyl 3-bromo-4-iodobenzoate

  • MDL number MFCD11520423
  • Cas Number : 386267-31-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerator
Physical Form White to light-red solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/51594.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/51594.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart