SY3H3D67D82F Synthonix Corporation

Ethyl 2,4-difluorobenzoate

  • MDL number MFCD00153149
  • Cas Number : 108928-00-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Clear liquid / Colour: Colorless - Yellow - Brown
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/51611.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/51611.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart