SY3H3D67D830 Synthonix Corporation

Ethyl 2,5-difluorobenzoate

  • MDL number MFCD06204339
  • Cas Number : 708-25-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerator
Physical Form Liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/51612.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/51612.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart