SY3H3D67D836 Synthonix Corporation

Ethyl 2-amino-6-fluoro-3-nitrobenzoate

  • Cas Number : 150368-37-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/51691.html

Safety Information

MSDS https://synthonix.com/51691.html
Availability details for
Quantity
  • Availability
  • Add to Cart