SY3H3D67D83D Synthonix Corporation

Ethyl 7-aminothieno[2,3-b]pyrazine-6-carboxylate

  • MDL number MFCD00096361
  • Cas Number : 56881-21-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/51733.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/51733.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart