SY3H3D67D844 Synthonix Corporation

Ethyl 4,6-dichloro-2-methylnicotinate

  • MDL number MFCD05666193
  • Cas Number : 686279-09-4
  • Molecular Weight : 234.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl 4,6-dichloro-2-methylnicotinate
Inchi Code 1S/C9H9Cl2NO2/c1-3-14-9(13)8-5(2)12-7(11)4-6(8)10/h4H,3H2,1-2H3
Inchi Key LNIMLSAAPISOEC-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. Refrigerator
Physical Form Colorless to Yellow Liquid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/51797.html

Safety Information

signalword Warning
pcodes P261,P280,P305,P351,P338
MSDS https://synthonix.com/51797.html
pictograms GHS07
hstatements H302,H315,H319,H332,H335
Availability details for
Quantity
  • Availability
  • Add to Cart