SY3H3D67D84C Synthonix Corporation

3-Ethoxy-4-hydroxy-5-nitrobenzaldehyde

  • MDL number MFCD00058678
  • Cas Number : 178686-24-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form White to off-white solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/51858.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/51858.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart