SY3H3D67D852 Synthonix Corporation

Ethyl 4-bromo-3-hydroxybenzoate

  • MDL number MFCD09038129
  • Cas Number : 33141-66-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerator
Physical Form White to off-white powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/51968.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/51968.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart