SY3H3D67D856 Synthonix Corporation

2-Ethoxybenzimidamide hydrochloride

  • MDL number MFCD05663676
  • Cas Number : 18637-00-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerator
Physical Form White to off-white powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/52131.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/52131.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart