SY3H3D67D86A Synthonix Corporation

Ethyl imidazo[1,2-a]pyridine-3-carboxylate

  • Cas Number : 123531-52-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/52317.html

Safety Information

MSDS https://synthonix.com/52317.html
Availability details for
Quantity
  • Availability
  • Add to Cart