SY3H3D67D86F Synthonix Corporation

Ethyl 2-aminobenzo[d]thiazole-6-carboxylate

  • MDL number MFCD00102724
  • Cas Number : 50850-93-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. refrigerator
Physical Form Crystal - Powder / Colour: Off white - yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/52329.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/52329.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart