SY3H3D67D870 Synthonix Corporation

Ethyl 2-aminobenzo[d]thiazole-7-carboxylate

  • MDL number MFCD13195422
  • Cas Number : 677304-89-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/52330.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/52330.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart