SY3H3D67D87B Synthonix Corporation

Ethyl 3-fluoro-6-iodo-2-methylbenzoate

  • MDL number MFCD28041189
  • Cas Number : 1417190-32-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/52445.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/52445.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart