SY3H3D67D87D Synthonix Corporation

Ethyl 4-formylbenzoate

  • Cas Number : 6287-86-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/52452.html

Safety Information

MSDS https://synthonix.com/52452.html
Availability details for
Quantity
  • Availability
  • Add to Cart