SY3H3D67D881 Synthonix Corporation

Ethyl 1-methyl-1H-benzo[d][1,2,3]triazole-5-carboxylate

  • MDL number MFCD04110946
  • Cas Number : 499785-52-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/52533.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/52533.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart