SY3H3D67D882 Synthonix Corporation

5-(4-Ethylphenyl)-1,3,4-thiadiazol-2-amine

  • MDL number MFCD02664104
  • Cas Number : 383130-78-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form White to light-yellow to yellow powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/52534.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/52534.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart