SY3H3D67D88B Synthonix Corporation

Ethyl 2-(2-chlorophenyl)acetate

  • MDL number MFCD09264019
  • Cas Number : 40061-54-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerator
Physical Form Pale-yellow to Yellow-brown Liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/52634.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/52634.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart