SY3H3D67D893 Synthonix Corporation

1-(3-Ethoxy-4-fluorophenyl)ethanone

  • MDL number MFCD18375404
  • Cas Number : 1378655-34-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerator
Physical Form Liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/52646.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/52646.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart