SY3H3D67D894 Synthonix Corporation

Ethyl 2-(2-iodophenyl)acetate

  • MDL number MFCD00099258
  • Cas Number : 90794-29-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/52649.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/52649.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart