SY3H3D67D89A Synthonix Corporation

Ethyl 5,7-dimethyl-[1,2,4]triazolo[1,5-a]pyrimidine-2-carboxylate

  • Cas Number : 114040-29-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/52716.html

Safety Information

MSDS https://synthonix.com/52716.html
Availability details for
Quantity
  • Availability
  • Add to Cart