SY3H3D67D8A2 Synthonix Corporation

Ethyl 2,4-dimethylnicotinate

  • MDL number MFCD00051962
  • Cas Number : 37669-78-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerator
Physical Form Liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/52941.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/52941.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart