SY3H3D67D8A9 Synthonix Corporation

Ethyl 4-amino-3-methoxybenzoate

  • MDL number MFCD14543019
  • Cas Number : 73368-41-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerator
Physical Form Off-white to brown powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/52978.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/52978.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart