SY3H3D67D8B3 Synthonix Corporation

Ethyl 4,5,6,7-tetrahydro-1H-indazole-5-carboxylate

  • MDL number MFCD21364708
  • Cas Number : 792848-34-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerator
Physical Form Pale-yellow to Yellow-brown Solid or Liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/53051.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/53051.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart