SY3H3D67D8B9 Synthonix Corporation

Ethyl 2-methyl-4-oxocyclohex-2-enecarboxylate

  • MDL number MFCD00001582
  • Cas Number : 487-51-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerator
Physical Form clear, liquid Colour: pale-yellow to dark yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/53099.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/53099.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart