SY3H3D67D8C1 Synthonix Corporation

5-(4-Ethylpiperazin-1-yl)pyrimidine

  • MDL number MFCD26743619
  • Cas Number : 894853-96-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form Liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/53200.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/53200.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart