SY3H3D67D8C3 Synthonix Corporation

Ethyl 2-(4,4-difluorocyclohexyl)acetate

  • MDL number MFCD18909256
  • Cas Number : 915213-54-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/53210.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/53210.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart