SY3H3D67D8C8 Synthonix Corporation

Ethyl 2-(2-azabicyclo[2.2.1]heptan-2-yl)acetate

  • MDL number MFCD14650178
  • Cas Number : 1250423-37-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/53231.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/53231.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart