SY3H3D67D8C9 Synthonix Corporation

Ethyl 6,8-dichlorooctanoate

  • MDL number MFCD01711127
  • Cas Number : 1070-64-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form liquid Colour: gold - Yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/53275.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/53275.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart