SY3H3D67D8CC Synthonix Corporation

ethyl 4-(hydroxymethyl)cyclohexane-1-carboxylate

  • Cas Number : 104802-52-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/53278.html

Safety Information

MSDS https://synthonix.com/53278.html
Availability details for
Quantity
  • Availability
  • Add to Cart