SY3H3D67D8CE Synthonix Corporation

(S)-Ethyl 2-acetamido-4-methylpentanoate

  • MDL number MFCD09952004
  • Cas Number : 4071-36-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form Liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/53293.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/53293.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart