SY3H3D67D8D2 Synthonix Corporation

5-Ethynyl-3-methyl-1H-indazole

  • MDL number MFCD14584652
  • Cas Number : 1093307-29-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form White to off-white solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/53632.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/53632.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart