SY3H3D67D8D6 Synthonix Corporation

Ethyl 4-bromothieno[2,3-c]pyridine-2-carboxylate

  • MDL number MFCD30188747
  • Cas Number : 1809004-78-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerator
Physical Form Light-yellow to brown powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/53754.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/53754.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart