SY3H3D67D8D9 Synthonix Corporation

Ethyl 4-chlorothieno[2,3-b]pyridine-5-carboxylate

  • MDL number MFCD09038145
  • Cas Number : 59713-58-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/53763.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/53763.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart