SY3H3D67D8DC Synthonix Corporation

4-Ethynyl-3-methylbenzoic acid

  • Cas Number : 1357281-69-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/53795.html

Safety Information

MSDS https://synthonix.com/53795.html
Availability details for
Quantity
  • Availability
  • Add to Cart