SY3H3D67D8E4 Synthonix Corporation

Ethyl 2-chloro-1H-benzo[d]imidazole-6-carboxylate

  • Cas Number : 857035-29-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/53857.html

Safety Information

MSDS https://synthonix.com/53857.html
Availability details for
Quantity
  • Availability
  • Add to Cart