SY3H3D67D8EC Synthonix Corporation

Ethyl 6-nitroimidazo[1,2-a]pyridine-2-carboxylate

  • Cas Number : 38923-08-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/53886.html

Safety Information

MSDS https://synthonix.com/53886.html
Availability details for
Quantity
  • Availability
  • Add to Cart