SY3H3D67D8EE Synthonix Corporation

Ethyl 3-cyanobenzoate

  • MDL number MFCD03086010
  • Cas Number : 2463-16-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. refrigerator
Physical Form White to off-white solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/53919.html

Safety Information

signalword Warning
pcodes P280
MSDS https://synthonix.com/53919.html
pictograms GHS07
hstatements H317, H319
Availability details for
Quantity
  • Availability
  • Add to Cart