SY3H3D67D8F4 Synthonix Corporation

Ethyl 3-oxo-2,3-dihydrofuro[2,3-b]pyridine-2-carboxylate

  • MDL number MFCD18074430
  • Cas Number : 1279893-97-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form White to off-white solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/53979.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/53979.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart