SY3H3D67D8F6 Synthonix Corporation

2-(Ethoxycarbonyl)-6-nitrobenzoic acid

  • MDL number MFCD01365751
  • Cas Number : 16533-45-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerator
Physical Form White to off-white solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/53985.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/53985.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart