SY3H3D67D8F7 Synthonix Corporation

Ethyl 2-(4-Chlorophenyl)-2,2-difluoroacetate

  • MDL number MFCD11007726
  • Cas Number : 130754-19-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form Liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/53999.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/53999.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart